Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NVS PAK1 C
GREENER_CHOICESDP

Catalog No. 61335 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-335 5 mg
61-332-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-335 Supplier Tocris Bioscience™ Supplier No. 6133/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Negative control of NVS PAK1 1

NVS PAK1 C is a negative control of NVS PAK1 1 (Cat. No. 6132), a potent and selective PAK1 inhibitor.

Specifications

Synonym (3S)-3-(2-Chloro-5-(2,2-difluoroethyl)-8-fluoro-5H-dibenzo[b,e][1,4]diazepin-11-ylamino)-N,N-dimethylpyrrolidine-1-carboxamide
Quantity 5 mg
Molecular Formula C22H23ClF3N5O
Formula Weight 465.91
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.