Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NVS-STG2
GREENER_CHOICESDP

Catalog No. 812010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
81-201-0 10 mg
81-205-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 81-201-0 Supplier Tocris Bioscience™ Supplier No. 8120/10
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

STING agonist, molecular glue that causes oligomerization of STING

NVS-STG2 is a STING agonist. It induces oligomerization of human STING by binding to pockets between adjacent STING dimer transmembrane domains, acting as a molecular glue. NVS-STG2 elicits potent STING-mediated immune responses in cells and produces antitumor activities in hSTING knock-in mice.

Specifications

Chemical Name or Material 4-((4-(2-(tert-Butyl)-5,5-dimethyl-1,3-dioxan-2-yl)benzyl)amino)-3-methoxybenzoic acid
CAS 3030588-01-0
Quantity 10 mg
Target STING Agonists
Molecular Formula C25H33NO5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.