Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NWD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140335 Supplier Invitrogen™ Supplier No. PA5140335
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ATVFLREIQD LHKHILEDCA LRMVDRLADG CLDTDAQNLL SSLKSHITDM HPGVLKTHRL PWSRDLVNPK NKTHACYLKE LGEQFVVRAN HQVLT Highest antigen sequence indentity to the following orthologs: Rat - 76%, Mouse - 73%.

NWD1 also named as NWD1 Ens or NACHT and WD repeat domain containing 1 is a 1564 amino acid protein, which contains 1 NACHT domain and 13 WD repeat. The molecular weight of NWD1 was detected by western blot. The result shows that the molecular weight of NWD1 is 77 kDa., but NWD1 is a 174 kDa protein according to UniProt,. The function of NWD1 is still non-known, meanwhile the molecular weight of NWD1 is still remained to be confirmed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NWD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NWD1
Gene Accession No. Q149M9
Gene Alias A230063L24Rik; NACHT and WD repeat domain containing 1; NACHT and WD repeat domain-containing protein 1; NACHT domain- and WD repeat-containing protein 1; Nwd1
Gene Symbols NWD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NWD1. Recombinant protein control fragment (Product #RP-109482).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.