Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NWSHPQFEK Rabbit, HRP, Polyclonal, GenScript™

Catalog No. 502120504
Encompass_Preferred
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-212-0504 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-212-0504 Supplier Genscript Corporation Supplier No. A0087540
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. Well-characterized antibodies for epitope tags consisting of short sequences have been widely used in the study of protein expression in various systems.The NWSHPQFEK, a nine-amino acid peptide that is capable of binding to the biotin pocket of streptavidin was engineered. The Rabbit anti NWSHPQFEK polyclonal antibody is developed in rabbit using a synthetic NWSHPQFEK peptide conjugated to KLH.This polyclonal antibody is highly purified from rabbit antiserum by immunoaffinity chromatography and then conjugated to horseradish peroxidase. The antibody is suitable for detecting fusion proteins that contain NWSHPQFEK.

NWSHPQFEK Polyclonal antibody specifically detects NWSHPQFEK in Tag samples. It is validated for ELISA, Western Blot

Specifications

Antigen NWSHPQFEK
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate HRP
Formulation PBS with 1% BSA and 0.01% thimerosal; pH 7.4
Host Species Rabbit
Immunogen Strep-tag II epitope peptide NWSHPQFEK conjugated to KLH.
Purification Method Affinity chromatography
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.