Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NWSHPQFEK Tag Monoclonal Antibody (5A9F9), THE™, GenScript™

Catalog No. 50273867
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-867 200 μg
50-273-866 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-273-867 Supplier Genscript Corporation Supplier No. A01732200
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. Streptavidin is a tetrameric protein purified from Streptomyces avidinii. It has wide used in numerous molecular biological protocols dues to its strong affinity for biotin. NWSHPQFEK (Strep tag II) is a nine amino acid peptide with high specificity and affinity towards streptavidin. NWSHPQFEK Tag Antibody is a useful tool in analysis and affinity purification of Strep tag II fusion proteins.

NWSHPQFEK Tag Monoclonal antibody specifically detects NWSHPQFEK Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen NWSHPQFEK Tag
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5A9F9
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Host Species Mouse
Immunogen NWSHPQFEK peptide conjugated to KLH
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.