Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NWSHPQFEK Tag Mouse, Biotin, Clone: 5A9F9, THE™, GenScript™

Catalog No. 50273875
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-273-875 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-273-875 Supplier Genscript Corporation Supplier No. A01737100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. Streptavidin is a tetrameric protein purified from Streptomyces avidinii. It has wide use in numerous molecular biological protocols dues to its strong affinity for biotin. NWSHPQFEK (Strep II tag) is a nine amino acid peptide with high specificity and affinity towards streptavidin.GenScript THE™ NWSHPQFEK Tag Antibody [Biotin], mAb, Mouse is THETM NWSHPQFEK Tag Antibody (A01732) conjugated with biotin. The conjugated antibody is suitable for detecting the expression level of NWSHPQFEK (Strep II tag) tagged fusion proteins.

NWSHPQFEK Tag Monoclonal antibody specifically detects NWSHPQFEK Tag in Tag samples. It is validated for ELISA, Western Blot

Specifications

Antigen NWSHPQFEK Tag
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5A9F9
Conjugate Biotin
Formulation PBS with 10mg/mL BSA and 0.02% sodium azide; pH 7.4
Host Species Mouse
Immunogen NWSHPQFEK peptide conjugated to KLH
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.