Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NXF1 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306455C
This item is not returnable.
View return policy
Description
NXF1 Polyclonal antibody specifically detects NXF1 in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
NXF1 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
10482 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 650 | |
DKFZp667O0311, Mex67, mRNA export factor TAP, nuclear RNA export factor 1, TAPMEX67, tip associating protein, Tip-associated protein, Tip-associating protein | |
Produced in rabbits immunized with E. coli-derived Human NXF1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction