Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NXF1 Monoclonal Antibody (2C2H1), Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 679141IG150UL


Description
NXF1 Monoclonal antibody specifically detects NXF1 in Human samples. It is validated for Western BlotSpecifications
NXF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MEX67; NXF1; TAP | |
Protein G | |
RUO | |
10482 | |
-20°C | |
Liquid |
Western Blot | |
2C2H1 | |
Unconjugated | |
Q9UBU9 | |
Mouse | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction