Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NXF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215049
Description
NXF2 Polyclonal specifically detects NXF2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
NXF2 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
CT39TAPL-2Cancer/testis antigen 39, FLJ20416, nuclear RNA export factor 2, NXF2B, TAP like protein 2, TAPL2, TAP-like protein 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
56001 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.79mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
Q9GZY0 | |
NXF2 | |
Recombinant protein encompassing a sequence within the center region of human NXF2. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction