Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104350 Supplier Invitrogen™ Supplier No. PA5104350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NXF2.

This gene is one of a family of nuclear RNA export factor genes. It encodes a protein that is involved in mRNA export, is located in the nucleoplasm, and is associated with the nuclear envelope. Alternative splicing seems to be a common mechanism in this gene family. Two variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXF2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NXF2
Gene Accession No. Q9GZY0
Gene Alias 4930455N06Rik; cancer/testis antigen 39; CT39; Nuclear RNA export factor 2; nuclear RNA export factor 5; NXF2; NXF2B; Nxf-b; RGD1559530; TAPL2; TAPL-2; TAP-like protein 2; t-complex protein 11 homolog; TCP11X2
Gene Symbols NXF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NXF2; NXF2B(Accession Q9GZY0), corresponding to amino acid residues Q26-L76.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308653, 56001, 83454
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.