Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NXF5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | NXF5 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15726020
![]() |
Novus Biologicals
NBP15726020UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157260
![]() |
Novus Biologicals
NBP157260 |
100 μL |
Each for $487.50
|
|
|||||
Description
NXF5 Polyclonal specifically detects NXF5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NXF5 | |
Polyclonal | |
Purified | |
RUO | |
nuclear RNA export factor 5, TAPL1, TAPL-1, TAP-like protein 1 | |
NXF5 | |
IgG | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Q9H1B4 | |
55998 | |
Synthetic peptides corresponding to NXF5 (nuclear RNA export factor 5) The peptide sequence was selected from the middle region of NXF5. Peptide sequence ITERNFPELLSLNLCNNKLYQLDGLSDITEKAPKVKTLNLSKNKLESAWE. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title