Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NXF5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112632
Description
NXF5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Could be involved in the export of mRNA from the nucleus to the cytoplasm. Could also have a role in polarized cytoplasmic transport and localization of mRNA in neurons.
Specifications
NXF5 | |
Polyclonal | |
Unconjugated | |
NXF5 | |
Nuclear RNA export factor 5; NXF5; TAPL1; TAPL-1; TAP-like protein 1 | |
Rabbit | |
Protein G | |
RUO | |
55998 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9H1B4 | |
NXF5 | |
Recombinant Human Nuclear RNA export factor 5 protein (1-200AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction