Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554825
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554825 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554825 Supplier Invitrogen™ Supplier No. PA554825
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PIAEKIIAKY KAKEEEAPLL FFVAGEDDMT DSLRDYTNLP EAAPLLTILD MSARAKYVMD VEEITPAIVE AFVNDFLAEK LKPEPI Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a member of the thioredoxin superfamily, a group of small, multifunctional redox-active proteins. Members of this family are characterized by a conserved active motif called the thioredoxin fold that catalyzes disulfide bond formation and isomerization. The encoded protein acts a redox-dependent regulator of the Wnt signaling pathway and is involved in cell growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXN
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NXN
Gene Accession No. P97346, Q6DKJ4
Gene Alias Gn25; l11Jus13; NRX; nucleoredoxin; nucleoredoxin 1; NXN; Protein Red-1; TRG-4
Gene Symbols NXN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NXN. Recombinant protein control fragment (Product #RP-93378).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18230, 360577, 64359
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.