Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NXNL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100361
Description
Antibody detects endogenous levels of total TXNL6.
Retinitis pigmentosa (RP) is a disease that leads to blindness by degeneration of cone photoreceptors. Rods produce factors required for cone viability. The protein encoded by this gene is one of those factors and is similar to a truncated form of thioredoxin. This gene has been proposed to have therapeutic value against RP.
Specifications
NXNL1 | |
Polyclonal | |
Unconjugated | |
Nxnl1 | |
A930031O08Rik; nucleoredoxin like 1; nucleoredoxin-like 1; nucleoredoxin-like protein 1; NXNL1; RdCVF; rod-derived cone viability factor; thioredoxin-like 6; thioredoxin-like protein 6; Txnl6 | |
Rabbit | |
Affinity chromatography | |
RUO | |
115861, 234404 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8VC33, Q96CM4 | |
Nxnl1 | |
A synthesized peptide derived from human NXNL1(Accession Q96CM4), corresponding to amino acid residues M1-A43. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction