Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NXNL2 Antibody (OTI2B11), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273134PEATT594
Description
NXNL2 Monoclonal specifically detects NXNL2 in Human, Mouse, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
NXNL2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
NXNL2 | |
Full length recombinant protein of human NXNL2 (NP_660326) produced in HEK293T cells. | |
0.1 mL | |
158046.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI2B11 | |
Flow Cytometry | |
C9orf121, chromosome 9 open reading frame 121, nucleoredoxin-like 2, nucleoredoxin-like protein 2, RDCVF2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Monkey, Primate | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction