Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXNL2 Monoclonal Antibody (OTI2H8)
GREENER_CHOICE

Catalog No. PIMA525134
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525134 Supplier Invitrogen™ Supplier No. MA525134
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NXNL2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

The specific function of NXNL2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXNL2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI2H8
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NXNL2
Gene Accession No. Q5VZ03
Gene Alias 4930519N16Rik; C9orf121; nucleoredoxin like 2; nucleoredoxin-like 2; nucleoredoxin-like protein 2; nucleoredoxin-like protein 2-like; Nxnl2; RdCVF2; rod-derived cone viability factor 2
Gene Symbols NXNL2
Host Species Mouse
Immunogen Full length human recombinant protein of NXNL2 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158046
Target Species Human, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.