Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXPE3 Monoclonal Antibody (4-E11)
GREENER_CHOICE

Catalog No. PIMA532894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532894 Supplier Invitrogen™ Supplier No. MA532894
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NXPE3 Monoclonal Antibody for ICC/IF, Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NXPE3
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4-E11
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene NXPE3
Gene Accession No. Q969Y0
Gene Alias FAM55C; family with sequence similarity 55, member C; MST115; MSTP115; neurexophilin and PC-esterase domain family member 3; neurexophilin and PC-esterase domain family, member 3; NXPE family member 3; NXPE3; Protein FAM55C; UNQ5791/PRO19596
Gene Symbols NXPE3
Host Species Mouse
Immunogen Recombinant protein with Human FAM55C aa 155-429.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91775
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.