Learn More
Invitrogen™ NXPH1 Recombinant Rabbit Monoclonal Antibody (021)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA531316
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Rat Neurexophilin-1/NXPH1.
NXPH1 is a secreted protein with a variable N-terminal domain, a highly conserved, N-glycosylated central domain, a short linker region, and a cysteine-rich C-terminal domain. This protein forms a very tight complex with alpha neurexins, a group of proteins that promote adhesion between dendrites and axons.
Specifications
NXPH1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q63366 | |
NXPH1 | |
Recombinant Rat Neurexophilin-1/NXPH1 protein (Met1-Gly271). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
ELISA | |
021 | |
Unconjugated | |
NXPH1 | |
C130005L03Rik; Nbla00697; neurexophilin 1; neurexophilin-1; Neurophilin; Nph1; NXPH1 | |
Rabbit | |
Protein A | |
RUO | |
25501 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.