Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXPH3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101710
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101710 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101710 Supplier Invitrogen™ Supplier No. PA5101710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NXPH3.

May be signaling molecules that resemble neuropeptides. Ligand for alpha-neurexins. Target protein is a member of a family of neuropeptide-like glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXPH3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NXPH3
Gene Accession No. O95157, Q91VX5, Q9Z2N5
Gene Alias KIAA1159; neurexophilin 3; neurexophilin-3; NPH3; Nxph3; UNQ687/PRO1327
Gene Symbols NXPH3
Host Species Rabbit
Immunogen A synthesized peptide derived from human NXPH3(Accession O95157), corresponding to amino acid residues F157-S207.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104079, 11248, 59315
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.