Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NXPH4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536496
Description
This antibody detects endogenous protein at a molecular weight of 33 kDa. Purity is >95% by SDS-PAGE.
Members of the Neurexophilin family are neuropeptide-like glycoproteins that undergo proteolytic processing after synthesis. As secreted proteins, Neurexophilins are thought to function as signaling molecules which specifically bind to target proteins, such as neurexin I-alpha (a protein that promotes adhesion between dendrites and axons), and are essential for proper neurotransmitter release. Neurexophilins contain five common domains including a variable N-terminal domain, a highly conserved N-glycosylated central domain, a short linker region and a cysteine-rich conserved C-terminal domain. Neurexophilin-4, also known as NXPH4 or NPH4, is a 308 amino acid secreted protein belonging to the neurexophilin family. Neurexophilin-4 is expressed in brain, spleen and testis.
Specifications
NXPH4 | |
Polyclonal | |
Unconjugated | |
NXPH4 | |
1110036M10Rik; AI851051; alternative protein NXPH4; neurexophilin 4; neurexophilin-4; Nph4; Nxph4; UNQ1928/PRO4403 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104080, 11247, 59316 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O95158, Q9Z2N4 | |
NXPH4 | |
Synthetic peptide corresponding to amino acids 221-270 of human Neurexophilin-4. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction