Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXT1 Mouse anti-Human, Mouse, Rat, Clone: 2C10A2, ProteinTech
SDP

Catalog No. 502239739 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9739 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9739 Supplier Proteintech Group Inc Supplier No. 676801IG150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aliquoting is unnecessary for -20°C storage.

NXT1 Monoclonal antibody specifically detects NXT1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen NXT1
Applications Western Blot
Classification Monoclonal
Clone 2C10A2
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene NXT1
Gene Accession No. Q9QZV9, Q9UKK6
Gene Alias 1110001N02Rik; MTR2; NTF2-like export factor 1; NTF2-related export protein 1; NTX2-like export factor1; nuclear transport factor 2 like export factor 1; nuclear transport factor 2-like export factor 1; NUTF-like export factor 1; NXT1; P15; Protein p15
Gene Symbols NXT1
Host Species Mouse
Immunogen NXT1 fusion protein Ag30039
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29107, 296219, 56488
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.