Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NXT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572140
Description
NXT1 Polyclonal Antibody for Western Blot
The protein encoded by this gene is located in the nuclear envelope. It has protein similarity to nuclear transport factor 2. This protein functions as a nuclear export factor in both RAN (Ras-related nuclear protein)- and CRM1 (required for chromosome region maintenance)-dependent pathways. It is found to stimulate the export of U1 snRNA in RAN- and CRM1-dependent pathways and the export of tRNA and mRNA in a CRM1-independent pathway. The encoded protein heterodimerizes with Tap protein and may regulate the ability of Tap protein to mediate nuclear mRNA export. The use of alternate polyadenylation sites has been found for this gene.
Specifications
NXT1 | |
Polyclonal | |
Unconjugated | |
NXT1 | |
1110001N02Rik; MTR2; NTF2-like export factor 1; NTF2-related export protein 1; NTX2-like export factor1; nuclear transport factor 2 like export factor 1; nuclear transport factor 2-like export factor 1; NUTF-like export factor 1; NXT1; P15; Protein p15 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
29107, 56488 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9QZV9, Q9UKK6 | |
NXT1 | |
KLH conjugated synthetic peptide between 85-111 amino acids from the C-terminal region of human NXT1. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction