Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NXT2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567222
Description
Immunogen sequence: KYRSHWSQGDR EGYQRRSNYY EGPHTSHSSP ADRTREEVVT PTLPEHTATR SQMATSLD Highest antigen sequence identity to the following orthologs - mouse 34%, rat 34%.
Regulator of protein export for NES-containing proteins. Also plays a role in mRNA nuclear export.
Specifications
NXT2 | |
Polyclonal | |
Unconjugated | |
NXT2 | |
6330587F24Rik; AI787442; BM-025; DC9; NTF2-related export protein 2; nuclear transport factor 2 like export factor 2; nuclear transport factor 2-like export factor 2; NXT2; P15-2; protein p15-2; RGD1560204 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55916 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPJ8 | |
NXT2 | |
Recombinant Human NXT2. Recombinant protein control fragment (Product #RP-107858). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction