Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NXT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589462 Supplier Invitrogen™ Supplier No. PA589462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLDALNNFFD TLPSSEFQVN MLDCQPVHEQ ATQSQTTVLV VTSGTVKFDG NKQHFFNQNF LLTAQSTPNN T; Positive Samples: Mouse testis, Rat testis; Cellular Location: Cytoplasm, Nucleus.

Regulator of protein export for NES-containing proteins. Also plays a role in mRNA nuclear export.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NXT2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NXT2
Gene Accession No. B2GV77, Q3UNA4, Q9NPJ8
Gene Alias 6330587F24Rik; AI787442; BM-025; DC9; NTF2-related export protein 2; nuclear transport factor 2 like export factor 2; nuclear transport factor 2-like export factor 2; NXT2; P15-2; protein p15-2; RGD1560204
Gene Symbols NXT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-180 of human NXT2 (NP_0611682).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237082, 315352, 55916
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.