Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NXT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727741 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7741 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7741 Supplier Proteintech Group Inc Supplier No. 121361AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Regulator of protein export for NES-containing proteins. Also plays a role in mRNA nuclear export.

Specifications

Antigen NXT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NXT2
Gene Accession No. B2GV77, Q3UNA4, Q9NPJ8
Gene Alias DC9, NTF2 related export protein 2, NXT2, P15 2, Protein p15 2
Gene Symbols Nxt2
Host Species Rabbit
Immunogen NXT2 Fusion Protein Ag2782
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 237082, 315352, 55916
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.