Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NY-ESO-1 Mouse anti-Human, Clone: D8.38, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC1151
Description
Detect NY-ESO-1, LAGE-1 using this mouse monoclonal Anti-NY-ESO-1 antibody, clone D8.38. Validated for use in Immunohistochemistry (Paraffin).
Specifications
NY-ESO-1 | |
Monoclonal | |
Please refer to lot specific datasheet. | |
CTAG1A,CTAG,CTAG1,ESO1,LAGE2,LAGE2A,CTAG1B,LAGE2B | |
A full length recombinant human NY-ESO-1 protein. | |
100 μL | |
Apoptosis and Cancer | |
NP_001318.1,NP_758965.2 | |
Human | |
Purified |
Immunohistochemistry (Paraffin) | |
D8.38 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone D8.38 specifically targets a sequence in the N-terminal region and detects Cancer/testis antigen 1 (NY-ESO1) in human testis | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction