Learn More
Invitrogen™ NY-ESO-1 Recombinant Rabbit Monoclonal Antibody (6U0N7)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA558010

Description
Immunogen sequence: MQAEGRGTGG STGDADGPGG PGIPDGPGGN AGGPGEAGAT GGRGPRGAGA ARASGPGGGA PRGPHGGAAS GLNGCCRCGA RGPESRLLEF YLAMPFATPM.
NY-ESO-1, a member of the CT (cancer/testis) family, is a tumor specific shared antigen with distinctive immunogenicity. The molecular basis of T cell-mediated antitumor immunity has been elucidated by the identification of a number of tumor antigens recognized by CD8+ T cells. Among these antigens, NY-ESO-1 is of particular interest because both the cytotoxic T lymphocyte and antibody have been shown to react with this antigen. The NY-ESO-1 has been detected in normal testis and in a range of human tumor types. NY-ESO-1 reactivity in testis is restricted germ cells, particularly in the spermatogonia.
Specifications
NY-ESO-1 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
P78358 | |
CTAG1A | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NY-ESO-1 0. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Western Blot | |
6U0N7 | |
Unconjugated | |
CTAG1A | |
Autoimmunogenic cancer/testis antigen NY-ESO-1; cancer/testis antigen 1; cancer/testis antigen 1A; cancer/testis antigen 1-A; cancer/testis antigen 6.1; CT6.1; CTAG; CTAG1; CTAG1A; CTAG1B; ESO1; l antigen family member 2; LAGE2; LAGE-2; LAGE2A; LAGE2B; New York Esophageal Squamous Cell Carcinoma 1; NY-ESO-1 | |
Rabbit | |
Protein A | |
RUO | |
246100 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.