Learn More
Invitrogen™ NYREN18 Monoclonal Antibody (OTI4H2)

Description
NYREN18 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | NYREN18 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI4H2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NUB1 |
| Gene Accession No. | Q9Y5A7 |
| Gene Alias | 4931404D21Rik; 6330412F12Rik; BS4; EGK_14356; NEDD8 ultimate buster 1; NEDD8 ultimate buster-1; negative regulator of ubiquitin like proteins 1; negative regulator of ubiquitin-like proteins 1; Nub1; NUB1L; NYREN18; NY-REN-18; NY-REN-18 antigen; Protein BS4; Renal carcinoma antigen NY-REN-18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.