Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NYREN18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599190 Supplier Invitrogen™ Supplier No. PA599190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NYREN18.

NYREN18 is a specific down-regulator of the NEDD8 conjugation system. It recruits NEDD8 and its conjugates to the proteasome for degradation. It also interacts with PSMD4/S5a, a member of the regulatory subunit of the 26S proteasome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NYREN18
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NUB1
Gene Accession No. P54729, Q9Y5A7
Gene Alias 4931404D21Rik; 6330412F12Rik; BS4; EGK_14356; NEDD8 ultimate buster 1; NEDD8 ultimate buster-1; negative regulator of ubiquitin like proteins 1; negative regulator of ubiquitin-like proteins 1; Nub1; NUB1L; NYREN18; NY-REN-18; NY-REN-18 antigen; Protein BS4; Renal carcinoma antigen NY-REN-18
Gene Symbols NUB1
Host Species Rabbit
Immunogen A synthesized peptide derived from human NUB1(Accession Q9Y5A7), corresponding to amino acid residues E581-N615.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51667, 53312
Target Species Human, Mouse, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.