Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-O-Acetyl-L-malic Anhydride 97.0+%, TCI America™
SDP

Catalog No. A110125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A110125G 25 g
A11015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A110125G Supplier TCI America Supplier No. A110125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59025-03-5
Molecular Formula C6H6O5
Molecular Weight (g/mol) 158.109
MDL Number MFCD00082708
InChI Key SSWJHSASZZAIAU-BYPYZUCNSA-N
Synonym s---2-acetoxysuccinic anhydride, --o-acetyl-l-malic anhydride, s-2-acetoxysuccinic anhydride, 2,5-furandione, 3-acetyloxy dihydro-, 3s, s-acetoxysuccinic anhydride, --o-acetylmalic anhydride, o-acetyl-l-malic anhydride, s-3-acetoxy-dihydro-2,5-furandione, 2,5-dioxotetrahydro-3-furanyl acetate #, s-2,5-dioxotetrahydrofuran-3-yl acetate
PubChem CID 11126528
IUPAC Name [(3S)-2,5-dioxooxolan-3-yl] acetate
SMILES CC(=O)OC1CC(=O)OC1=O

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 25 g
Formula Weight 158.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-O-Acetyl-L-malic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.