Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Allylhydroxylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A14491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14491G 1 g
A14495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14491G Supplier TCI America Supplier No. A14491G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38945-21-0
Molecular Formula C3H8ClNO
Molecular Weight (g/mol) 109.553
MDL Number MFCD00012957
InChI Key XIQUJVRFXPBMHS-UHFFFAOYSA-N
PubChem CID 3084724
IUPAC Name O-prop-2-enylhydroxylamine;hydrochloride
SMILES C=CCON.Cl

Specifications

Melting Point 170°C
Color White-Yellow
Quantity 1 g
Formula Weight 109.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material O-Allylhydroxylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.