Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Anisaldehyde, 98%

Catalog No. p-60934
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle

CAS: 135-02-4 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-02-4
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00003308
InChI Key PKZJLOCLABXVMC-UHFFFAOYSA-N
Synonym o-anisaldehyde, benzaldehyde, 2-methoxy, 2-anisaldehyde, o-methoxybenzaldehyde, 6-methoxybenzaldehyde, salicylaldehyde methyl ether, 2-methoxybenzenecarboxaldehyde, benzaldehyde, o-methoxy, 2-methoxyphenylformaldehyde, formylanisole, o
PubChem CID 8658
IUPAC Name 2-methoxybenzaldehyde
SMILES COC1=CC=CC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34.0°C to 40.0°C
Color Brown to Yellow
Boiling Point 238.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5608
Linear Formula CH3OC6H4CHO
Quantity 5 g
Beilstein 08, IV, 180
Merck Index 10, 5866
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 136.15
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material o-Anisaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 205-171-7
RTECSNumber BZ2610000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.