Learn More
o-Benzoic sulfimide, 98+%
Description
o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
o-Benzoic sulfimide is used as a pharmaceutic flavor aid. It is used in high performance liquid chromatographic method for the simultaneous separation and determination of acesulfame potassium, saccharine and aspartame.
Solubility
Soluble in water (3.3 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 227°C to 230°C |
Density | 0.828 |
Quantity | 2500 g |
Beilstein | 6888 |
Merck Index | 14,8311 |
Solubility Information | Soluble in water (3.3g/L at 20°C). |
Formula Weight | 183.18 |
Percent Purity | ≥98% |
Chemical Name or Material | o-Benzoic sulfimide |
Safety and Handling
EINECSNumber | 201-321-0 |
RTECSNumber | DE4200000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.