Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzoyl-N-methylhydroxylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. B32391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32391G 1 g
B32395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32391G Supplier TCI America Supplier No. B32391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27130-46-7
Molecular Formula C8H10ClNO2
Molecular Weight (g/mol) 187.623
InChI Key FTCDQNQIGUCFSA-UHFFFAOYSA-N
PubChem CID 11615307
IUPAC Name methylamino benzoate;hydrochloride
SMILES CNOC(=O)C1=CC=CC=C1.Cl

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 1 g
Formula Weight 187.62
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material O-Benzoyl-N-methylhydroxylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.