Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Benzyl-L-serine, 99%

Catalog No. p-7026409
Click to view available options
Quantity:
5 g
25 g

CAS: 4726-96-9 | C10H13NO3 | 195.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4726-96-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.218
MDL Number MFCD00065937
InChI Key IDGQXGPQOGUGIX-VIFPVBQESA-N
Synonym o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine
PubChem CID 78457
IUPAC Name (2S)-2-amino-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)N

Specifications

Melting Point 213°C (decomposition)
Beilstein 2114846
Quantity 5 g
Formula Weight 195.22
Optical Rotation +7° (c=1 in 1N HCl)
Percent Purity 99%
Chemical Name or Material O-Benzyl-L-serine
EINECSNumber 225-220-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.