Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresol, 98+%

Catalog No. AAA123070P
Click to view available options
Quantity:
500 g
2000 g

CAS: 95-48-7 | C7H8O | 108.14 g/mol

o-Cresol is mainly used as a precursor to other compounds. Kolbe-Schmitt carboxylation gives o-cresotinic acid, a pharmaceutical intermediate. The muscle relaxant Mephenesin is an ether derived from o-cresol. Used as adhesives and sealant chemicals, fuels and fuel additives, intermediates, Solvents, Paints and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Cresol is mainly used as a precursor to other compounds. Kolbe-Schmitt carboxylation gives o-cresotinic acid, a pharmaceutical intermediate. The muscle relaxant Mephenesin is an ether derived from o-cresol. Used as adhesives and sealant chemicals, fuels and fuel additives, intermediates, Solvents, Paints and coatings.

Solubility
Soluble in chloroform, ether. Insoluble in water but soluble in about 40 parts water. Soluble in carbon tetrachloride

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-48-7
Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00002226
InChI Key QWVGKYWNOKOFNN-UHFFFAOYSA-N
Synonym o-cresol, 2-hydroxytoluene, o-methylphenol, 2-cresol, phenol, 2-methyl, orthocresol, o-cresylic acid, o-oxytoluene, o-toluol, o-hydroxytoluene
PubChem CID 335
ChEBI CHEBI:28054
IUPAC Name 2-methylphenol
SMILES CC1=CC=CC=C1O

Specifications

Melting Point 30°C to 34°C
Density 1.048
Boiling Point 190°C to 192°C
Flash Point 81°C (177°F)
Odor Phenol-like
Quantity 2000 g
UN Number UN3455
Beilstein 506917
Sensitivity Light sensitive
Merck Index 14,2579
Solubility Information Soluble in chloroform,ether. Insoluble in water but soluble in about 40 parts water. Soluble in carbon tetrachloride
Formula Weight 108.14
Percent Purity ≥98%
Chemical Name or Material o-Cresol
Show More Show Less
Hazard Category H227-H301+H311-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CRESOLS, SOLID
EINECSNumber 202-423-8
RTECSNumber GO6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.