Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresolphthalein (0.04% in ca. 95% Ethanol) [for pH Determination], TCI America™
SDP

Catalog No. C0218100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0218100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C0218100ML Supplier TCI America Supplier No. C0218100ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 596-27-0
Molecular Formula C22H18O4
Molecular Weight (g/mol) 346.38
MDL Number MFCD00005912
InChI Key CPBJMKMKNCRKQB-UHFFFAOYSA-N
Synonym o-cresolphthalein, cresolphthalein, 3,3'-dimethylphenolphthalein, 1 3h-isobenzofuranone, 3,3-bis 4-hydroxy-3-methylphenyl, 3',3-dimethylphenolphthalein, o-cresolphalein, 3,3-bis 4-hydroxy-3-methylphenyl isobenzofuran-1 3h-one, unii-xou2d9049g, phenolphthalein, 3',3-dimethyl, 3,3-bis 4-hydroxy-m-tolyl phthalide
PubChem CID 68995
IUPAC Name 3,3-bis(4-hydroxy-3-methylphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES CC1=CC(=CC=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

Color Colorless
pH 8.2-9.8
Quantity 100 mL
UN Number 1170
Formula Weight 346.38
Physical Form Liquid
Chemical Name or Material o-Cresolphthalein (0.04% in ca. 95% Ethanol) [for pH Determination]
RTECSNumber SM8390000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.