Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Cresolphthalein, 0.2% Alcoholic, Ricca Chemical
SDP

Catalog No. R2380000120 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R2380000120 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R2380000120 Supplier Ricca Chemical Company Supplier No. R2380000120A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 596-27-0 , 64-17-5 , 67-63-0 , 67-56-1
Molecular Formula C22H18O4
Molecular Weight (g/mol) Mixture
MDL Number MFCD00005912
InChI Key CPBJMKMKNCRKQB-UHFFFAOYSA-N
PubChem CID 68995
IUPAC Name 3,3-bis(4-hydroxy-3-methylphenyl)-1,3-dihydro-2-benzofuran-1-one
SMILES CC1=CC(=CC=C1O)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C(C)=C1

Specifications

CAS Min % 0.0, 88.0, 4.5, 4.0
CAS Max % 0.0, 89.0, 5.5, 5.0
Percent Purity 0.2% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.2% Alcoholic
Odor Odorless
Packaging Natural Poly Bottle
Color Colorless to Brown
Quantity 120 mL
Chemical Name or Material o-Cresolphthalein
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.