Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Formyl-D-mandeloyl Chloride 98.0+%, TCI America™
SDP

Catalog No. F072125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 29169-64-0
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.60
MDL Number MFCD00799229
InChI Key ZNLABNPTWSKGDX-UHFFFAOYNA-N
Synonym (R)-O-Formylmandeloyl Chloride
PubChem CID 11830408
IUPAC Name 2-chloro-2-oxo-1-phenylethyl formate
SMILES ClC(=O)C(OC=O)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 221°C
Quantity 25 g
UN Number 3265
Formula Weight 198.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material O-Formyl-D-mandeloyl Chloride
EINECSNumber (3)-4246
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.