Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-GlcNAc Antibody (9H6), Novus Biologicals™
SDP

Catalog No. p-200080965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259385 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259385 Supplier Novus Biologicals Supplier No. NBP259385
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

O-GlcNAc Monoclonal specifically detects O-GlcNAc in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen O-GlcNAc
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 9H6
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:25, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Gene Alias GlcNAc, O-linked N-acetylglucosamine
Host Species Mouse
Immunogen Synthetic B-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH).
Purification Method PEG purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Specific for B-O-Linked N-Acetylglucosamine modified proteins. Does not cross-react with N-Acetylgalactosamine, Glucose, or Galactose modified proteins.
Target Species Non-species specific
Content And Storage Store at -20C.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.