Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-GlcNAc Antibody (RL2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013965 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013965 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013965 Supplier Novus Biologicals Supplier No. NBP280892
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

O-GlcNAc Monoclonal specifically detects O-GlcNAc in Human, Mouse, Rat, Porcine, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Flow (Intracellular), CyTOF-ready.

Specifications

Antigen O-GlcNAc
Applications Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone RL2
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot : 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 20404350), Flow Cytometry : 1:10 - 1:1000, ELISA 1:100 - 1:2000,. Use reported in scientific literature (PMID 12029848), Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation : 1:10 - 1:500, Immunohistochemistry-Paraffin : 1:200, Dot Blot : 1:800, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, CyTOF-ready
Formulation PBS
Gene Alias GlcNAc, O-linked N-acetylglucosamine
Host Species Mouse
Immunogen Pore complex-lamina fraction purified from rat liver nuclear envelopes.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Porcine reactivity reported in scientific literature (PMID: 26004176). Xenopus reactivity reported in scientific literature (PMID: 17329255). Please note that this antibody is reactive to Mouse and derived from the same host, Mouse. Additional Mouse on Mouse blocking steps may be required for IHC and ICC experiments. Please contact Technical Support for more information.
Target Species Human, Mouse, Rat, Pig, Bovine, Drosophila, Fish, Hamster, Primate, Virus, Xenopus species
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.