Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
O-GlcNAc Monoclonal specifically detects O-GlcNAc in Bovine, Drosophila, Fish, Hamster, Human, Mouse, Pig, Primate, Rat, Virus, Xenopus species samples. It is validated for ChIP Assay, Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Specifications
Specifications
| Antigen | O-GlcNAc |
| Applications | ChIP Assay, Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | RL2 |
| Concentration | 1.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Chromatin Immunoprecipitation 1:10 - 1:500. Use reported in scientific literature (PMID 20404350), Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000. Use reported in scientific literature (PMID 12029848), Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200, Dot Blot 1:800, Chromatin Immunoprecipitation (ChIP) 1:10-1:500 |
| Gene Alias | GlcNAc, O-linked N-acetylglucosamine |
| Host Species | Mouse |
| Immunogen | Pore complex-lamina fraction purified from rat liver nuclear envelopes. |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
