Learn More
O-GlcNAc Mouse, Alkaline Phosphatase, Clone: 9H6, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB19217
Description
O-GlcNAc Monoclonal antibody specifically detects O-GlcNAc in samples. It is validated for WB, ELISA, ICC, IF.Specifications
O-GlcNAc | |
Monoclonal | |
Alkaline Phosphatase (AP) | |
Synthetic β-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH) | |
Primary | |
Liquid |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
9H6 | |
Mouse | |
100 μg | |
Store at -20°C | |
IgM |