Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-GlcNAc Mouse, PerCP, Clone: 9H6, Abnova™

Catalog No. 89338723 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-338-723 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-338-723 Supplier Abnova Corporation Supplier No. MAB19219
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

O-GlcNAc Monoclonal antibody specifically detects O-GlcNAc in samples. It is validated for WB, ELISA, ICC, IF.

Specifications

Antigen O-GlcNAc
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 9H6
Conjugate PerCP
Host Species Mouse
Immunogen Synthetic β-O-Linked N-Acetylglucosamine conjugated to Keyhole Limpet Hemocyanin (KLH)
Quantity 100 μg
Primary or Secondary Primary
Content And Storage Store at -20°C
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.