Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
O-GlcNAc Transferase p110 subunit Antibody (2037), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243755
Description
Ensure accurate, reproducible results in Western Blot
O-GlcNAc Transferase p110 subunit Monoclonal specifically detects O-GlcNAc Transferase p110 subunit in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, CyTOF-reported.Specifications
O-GlcNAc Transferase p110 subunit | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
OGT | |
Recombinant protein encompassing a sequence within the center region of human O-GlcNAc transferase. The exact sequence is proprietary. | |
0.1 mL | |
Amino Acids Drugs and other small molecules, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers | |
8473 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence, CyTOF | |
2037 | |
Western Blot 1:500 - 1:3000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, CyTOF-reported | |
EC 2.4.1, EC 2.4.1.255 FLJ23071, HRNT1, MGC22921, O-GLCNAC, O-GlcNAc transferase p110 subunit, O-GlcNAc transferase subunit p110, O-linked N-acetylglucosamine (GlcNAc) transferase(UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase), O-linked N-acetylglucosamine transferase 110 kDa subunit, UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDasubunit, uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyltransferase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction