Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
O-GlcNAcase/OGA/MGEA5 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232233JF549
Description
O-GlcNAcase/OGA/MGEA5 Polyclonal antibody specifically detects O-GlcNAcase/OGA/MGEA5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
O-GlcNAcase/OGA/MGEA5 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
bifunctional protein NCOAT, FLJ11229, HEXC, hyaluronidase in meningioma, KIAA0679, MEA5FLJ23355, meningioma expressed antigen 5 (hyaluronidase), Meningioma-expressed antigen 5, NCOAT, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, OGA, O-GlcNAcase | |
The immunogen this antibody was made to, maps to a region between residue 500 to 550 of human Meningioma Expressed Antigen 5 using the numbering given in entry NP_036347.1 (GeneID 10724). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
10724 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction