Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Methyl-L-tyrosine, 98%

Catalog No. aah6309606
Click to view available options
Quantity:
1 g
5 g

CAS: 6230-11-1 | C5H8O5 | 148.11 g/mol

O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-Methyl-L-tyrosine, is used as a fine chemical intermediate.

Solubility
Very soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6230-11-1
MDL Number MFCD00002604

Specifications

Melting Point 259°C to 261°C (decomposition)
Molecular Formula C10H13NO3
Beilstein 2212726
Synonym 4-methoxy-l-phenylalanine, o-methyl-l-tyrosine, h-tyr me-oh, o-methyltyrosine, s-2-amino-3-4-methoxyphenyl propanoic acid, p-methoxy-l-phenylalanine, 2s-2-amino-3-4-methoxyphenyl propanoic acid, tyrosine, o-methyl, l-4-methoxyphenylalanine, h-4-methoxy-phe-oh
Solubility Information Very soluble in water.
InChI Key GEYBMYRBIABFTA-IMWMWJONNA-N
SMILES COC1=CC=C(C[C@H](N)C(O)=O)C=C1
IUPAC Name (2S)-2-amino-3-(4-methoxyphenyl)propanoic acid
Quantity 5 g
Molecular Weight (g/mol) 195.22
PubChem CID 2723935
Formula Weight 195.22
Percent Purity 98%
Physical Form Solid
Chemical Name or Material O-Methyl-L-tyrosine
Show More Show Less
EINECSNumber 228-333-9
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.