Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-Methylisourea hemisulfate, 94%

Catalog No. p-164486
Click to view available options
Quantity:
25 g
250 g
1 kg

CAS: 52328-05-9 | 0·5 H2SO4 | 123.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52328-05-9
Molecular Formula 0·5 H2SO4
Molecular Weight (g/mol) 123.12
MDL Number MFCD00040594
InChI Key QSCPQKVWSNUJLJ-UHFFFAOYSA-N
Synonym o-methylisourea hemisulfate, 2-methylisouronium sulfate, o-methylisourea hemisulfate salt, bis methoxymethanimidamide ; sulfuric acid, carbamimidic acid, methyl ester, sulfate 2:1, omi™, o-methylpseudourea sulfate, 0-methylisourea hemisulfate, o-methylisourea hemisulphate, bis 2-methylisouronium sulphate
PubChem CID 11357018
IUPAC Name methyl carbamimidate;sulfuric acid
SMILES COC(=N)N.COC(=N)N.OS(=O)(=O)O

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 147°C to 153°C
Assay Percent Range 94%
Packaging Plastic bottle
Linear Formula 0·5H2SO4
Quantity 25 g
Solubility Information Solubility in water: 1000g/L water (20°C)
Formula Weight 123.12
Percent Purity 94%
Chemical Name or Material O-Methylisourea hemisulfate, 94%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 257-851-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.