Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O,N,N'-Triisopropylisourea 98.0+%, TCI America™
SDP

Catalog No. p-7137590
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 63460-32-2
Molecular Formula C10H22N2O
Molecular Weight (g/mol) 186.299
InChI Key RHNDDRWPYPWKNW-UHFFFAOYSA-N
PubChem CID 11332901
IUPAC Name propan-2-yl N,N'-di(propan-2-yl)carbamimidate
SMILES CC(C)NC(=NC(C)C)OC(C)C

Specifications

Color Colorless
Boiling Point 67°C
Quantity 5 mL
Synonym Isopropyl N,N′C-Diisopropylcarbamimidate, N,N′C-Diisopropylcarbamimidic Acid Isopropyl Ester
Formula Weight 186.30
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material O,N,N′-Triisopropylisourea
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.