Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

o-Phenylenediacetonitrile, 99+%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
613-73-0
C10H8N2
156.19
MFCD00001905
FWPFXBANOKKNBR-UHFFFAOYSA-N
1,2-phenylenediacetonitrile,o-phenylenediacetonitrile,1,2-bis cyanomethyl benzene,o-xylylene dicyanide,2-2-cyanomethyl phenyl acetonitrile,o-benzenediacetonitrile,2,2'-1,2-phenylene diacetonitrile,o-bis cyanomethyl benzene,1,2-benzenediacetonitrile,o-benzendiacetonitrile
69180
2-[2-(cyanomethyl)phenyl]acetonitrile
C1=CC=C(C(=C1)CC#N)CC#N
613-73-0
C10H8N2
156.19
MFCD00001905
FWPFXBANOKKNBR-UHFFFAOYSA-N
1,2-phenylenediacetonitrile,o-phenylenediacetonitrile,1,2-bis cyanomethyl benzene,o-xylylene dicyanide,2-2-cyanomethyl phenyl acetonitrile,o-benzenediacetonitrile,2,2'-1,2-phenylene diacetonitrile,o-bis cyanomethyl benzene,1,2-benzenediacetonitrile,o-benzendiacetonitrile
69180
2-[2-(cyanomethyl)phenyl]acetonitrile
C1=CC=C(C(=C1)CC#N)CC#N
Melting Point 58°C to 61°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 10g
Color Undesignated
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99% min. (GC)
Linear Formula C6H4(CH2CN)2
Beilstein 09, 874
Solubility Information Solubility in water: insoluble
Formula Weight 156.19
Physical Form Crystals, Crystalline Powder and/or Chunks
Percent Purity 99+%
Chemical Name or Material o-Phenylenediacetonitrile, 99+%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-351-3